missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TREM2 Antibody (08), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21453APC
290.70 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
TREM2 Monoclonal antibody specifically detects TREM2 in Human samples. It is validated for ELISA, ELISA (Matched Antibody Pair)
Specifications
| TREM2 | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, ELISA Detection (Matched Antibody Pair) | |
| 08 | |
| ELISA, ELISA Detection (Matched Antibody Pair) | |
| Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human TREM2 (Accession#: NP_061838.1; Met1-Ser174). | |
| 0.1 mL | |
| Alzheimers Research, Immunology, Inflammation, Innate Immunity, Neuroscience | |
| 54209 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction