missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TREM1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11791-1-AP-20UL
This item is not returnable.
View return policy
Description
TREM1 (triggering receptor expressed on myeloid cells 1) is a receptor belonging to the Ig superfamily that amplifies neutrophil and monocyte-mediated inflammatory responses triggered by bacterial and fungal infections by stimulating release of pro-inflammatory chemokines and cytokines, as well as increased surface expression of cell activation markers. It is thought that TREM1 other related TREM proteins such as TREM2 act to modulate the inflammatory response. TREM1 has also been linked to inflammatory bowel disease resulting in enhanced production of TNF, IL-6, IL-8, and MCP-1.Specifications
| TREM1 | |
| Polyclonal | |
| Unconjugated | |
| TREM1 | |
| CD354, TREM 1, TREM1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 54210, 58217 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Western Blot, Flow Cytometry, Immunocytochemistry | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9JKE2, Q9NP99 | |
| TREM1 | |
| TREM1 Fusion Protein Ag2418 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction