missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TREM1 Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA547090
This item is not returnable.
View return policy
Description
In sandwich ELISAs, less than 0.2% cross-reactivity with recombinant human (rh) TREM-1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
TREM1 (triggering receptor expressed on myeloid cells 1) is a receptor belonging to the Ig superfamily that amplifies neutrophil and monocyte-mediated inflammatory responses triggered by bacterial and fungal infections by stimulating release of pro-inflammatory chemokines and cytokines, as well as increased surface expression of cell activation markers. It is thought that TREM1 other related TREM proteins such as TREM2 act to modulate the inflammatory response. TREM1 has also been linked to inflammatory bowel disease resulting in enhanced production of TNF, IL-6, IL-8, and MCP-1.
Specifications
| TREM1 | |
| Polyclonal | |
| Unconjugated | |
| TREM1 | |
| CD354; sCD354; soluble CD354; soluble TREM 1; soluble TREM1; sTREM 1; sTREM1; TREM; Trem1; TREM-1; triggering receptor expressed in monocytes 1; triggering receptor expressed on monocytes 1; Triggering receptor expressed on myeloid cells; Triggering receptor expressed on myeloid cells 1 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 58217 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| ELISA, Flow Cytometry, Functional Assay, Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q9JKE2 | |
| TREM1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse TREM-1 Ala21-Ser202. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction