missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TREM1/CD64 Bispecific Recombinant Mouse Monoclonal Antibody (SAA2036)
Mouse Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA555043
This item is not returnable.
View return policy
Description
Compatible with Direct ELISA.
TREM1/CD64 enhances immune response by amplifying inflammation and promoting phagocytosis through activation of myeloid cells.
Specifications
| TREM1/CD64 Bispecific | |
| Recombinant Monoclonal | |
| 1.6 mg/mL | |
| PBS with no preservative | |
| Mouse | |
| Metal-chelate chromatography | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Neutralization | |
| SAA2036 | |
| Unconjugated | |
| MCP; MIC10; TREM1CD46 | |
| Recombinant Human TREM1 and CD46. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction