Learn More
TREM1 Antibody (L5-B8.2A12.3A12) [Janelia Fluor™ 635], Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP1-40907JF635
Product Code. 30171437
  • £509.00 / 0.25mL
Description
TREM1 Monoclonal antibody specifically detects TREM1 in Mouse samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Frozen),CyTOF,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TREM1
Monoclonal
Janelia Fluor 635
CD354 antigen, TREM-1Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1
Mouse TREM-1 Fc fusion protein
0.25 mL
Signal Transduction
54210
Store at 4°C in the dark.
IgG2a
Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Frozen), CyTOF, Immunocytochemistry/Immunofluorescence
L5-B8.2A12.3A12
50mM Sodium Borate
Rat
Protein G purified
RUO
Primary
Mouse
Purified