Learn More
TREM1 Antibody (CL7111) [Alexa Fluor™ 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-76531AF594
Product Code. 30308421
  • £558.00 / 0.10mL
Description
TREM1 Monoclonal antibody specifically detects TREM1 in Human samples. It is validated for Western Blot,KnockDown
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TREM1
Monoclonal
Alexa Fluor 594
CD354 antigen, TREM-1Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1
This antibody was developed using a synthetic peptide derived from Q9NP99, with the exact immunogen sequence remaining proprietary.
0.1 mL
Signal Transduction
54210
Store at 4⪚C in the dark.
IgG1
Western Blot, KnockDown
CL7111
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified