missing translation for 'onlineSavingsMsg'
Learn More

Trelagliptin, MedChemExpress

Product Code. p-200097876
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16492587

Brand: Medchem Express HY15408

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Trelagliptin (SYR-472) is a potent, orally active and highly selective DPP-4 inhibitor with an IC50 of 4 nM. Trelagliptin succinate improves glycemic control in vivo and can be used for the study of type 2 diabetes mellitus (T2DM).

Trelagliptin (SYR-472) is a potent, orally active and highly selective DPP-4 inhibitor with an IC50 of 4 nM. Trelagliptin succinate improves glycemic control in vivo and can be used for the study of type 2 diabetes mellitus (T2DM).

Specifications

Chemical Name or Material Trelagliptin
CAS 865759-25-7
Color White
Purity Grade Notes Research
Molecular Formula C18H20FN5O2
Quantity 50 mg
Synonym SYR-472
Solubility Information DMSO : ≥ 50 mg/mL (139.91 mM)
SMILES N#CC1=CC=C(F)C=C1CN(C(N2C)=O)C(N3C[C@H](N)CCC3)=CC2=O
Molecular Weight (g/mol) 357.38
Formula Weight 357.38
Percent Purity 99.8%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.