missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSF Progressive Safety™ TREKKER™ Safety Trainer
Click to view available options
:
Size: 7
Description
- All safety footwear is designed with both comfort and durability in mind
- Conforms to EN ISO 20345:2011 for 200 joules protection
- Black EVA/grey rubber outsole
- Black stitching with reflective side detail
- Black lace, 4 black/grey woven tabs with 1 black eyelet to top
- Black sandwich mesh lining
- Steel toe cap with composite midsole for weight and flexibility
Specifications
Specifications
| Certifications/Compliance | EN ISO 20345:2011 |
| Color | Black |
| For Use With (Application) | Hazardous conditions |
| Size | 7 |
| Style | Trainer |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction