missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TREK 1 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-41535F
This item is not returnable.
View return policy
Description
TREK 1 Polyclonal specifically detects TREK 1 in Human, Mouse, Bovine samples. It is validated for Immunohistochemistry (Negative), ICC/IF (Negative).
Specifications
| TREK 1 | |
| Polyclonal | |
| Immunohistochemistry (Negative), ICC/IF (Negative) | |
| O95069 | |
| KCNK2 | |
| A synthetic peptide made to an N-terminal region of human TREK 1 (within residues 1-100). [Swiss-Prot# Q9NRT2] | |
| 0.1 mL | |
| Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 3776 | |
| Human, Mouse, Bovine | |
| IgG |
| Immunohistochemistry, Immunocytochemistry | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| MGC126742, MGC126744, potassium channel, subfamily K, member 2, subfamily K, member 2, tandem-pore-domain potassium channel TREK-1, TREK-1 K(+) channel subunit, TWIK-related potassium channel 1, two-pore potassium channel 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, mouse, and cow. 88% sequence identity with rat protein. | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction