Learn More
Tregalizumab Recombinant Human Monoclonal Antibody, Invitrogen™
Human Recombinant Monoclonal Antibody
Brand: Invitrogen MA541937
Additional Details : Weight : 0.01000kg
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Tregalizumab has been used in trials studying the treatment of Rheumatoid Arthritis.Specifications
Tregalizumab | |
Recombinant Monoclonal | |
Unconjugated | |
hBF5; hB-F5; BT-061 | |
Human CD4 | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
ELISA, SDS-Page | |
0.76 mg/mL | |
PBS with no preservative; pH 7.5 | |
Human | |
Protein A | |
RUO | |
Chemical | |
Antibody | |
IgG1 κ |