missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Treacher Collins syndrome protein Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-03783-20UL
This item is not returnable.
View return policy
Description
Treacher Collins syndrome protein Polyclonal specifically detects Treacher Collins syndrome protein in Human samples. It is validated for Western Blot.
Specifications
| Treacher Collins syndrome protein | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| MFD1, nucleolar trafficking phosphoprotein, TCS1, Treacher Collins syndrome protein, Treacher Collins-Franceschetti syndrome 1, treacle, treacle protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 659-958 of human Treacher Collins syndrome protein (NP_001008657.1). | |
| 20 μL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 6949 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction