missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Treacher Collins syndrome protein Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Treacher Collins syndrome protein Polyclonal specifically detects Treacher Collins syndrome protein in Human, Mouse samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | Treacher Collins syndrome protein |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | MFD1, nucleolar trafficking phosphoprotein, TCS1, Treacher Collins syndrome protein, Treacher Collins-Franceschetti syndrome 1, treacle, treacle protein |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 659-958 of human Treacher Collins syndrome protein (NP_001008657.1). |
| Purification Method | Affinity purified |
| Quantity | 100 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?