missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals Treacher Collins syndrome protein Antibody (8H3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00006949-M02
This item is not returnable.
View return policy
Description
Treacher Collins syndrome protein Monoclonal antibody specifically detects Treacher Collins syndrome protein in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| Treacher Collins syndrome protein | |
| Monoclonal | |
| Unconjugated | |
| NP_001008657 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry | |
| 8H3 | |
| In 1x PBS, pH 7.4 | |
| MFD1, nucleolar trafficking phosphoprotein, TCS1, Treacher Collins syndrome protein, Treacher Collins-Franceschetti syndrome 1, treacle, treacle protein | |
| TCOF1 (NP_001008657, 2 a.a. ~ 82 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. AEARKRRELLPLIYHHLLRAGYVRAAREVKEQSGQKCFLAQPVTLLDIYTHWQQTSELGRKRKAEEDAALQAKKTRVSDPI | |
| 0.1 mg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 6949 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction