missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRBP Mouse anti-Human, Clone: PCRP-TARBP2-1E5, Novus Biologicals™
Mouse Monoclonal Antibody
£396.00
Specifications
| Antigen | TRBP |
|---|---|
| Clone | PCRP-TARBP2-1E5 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Protein Array |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence, Peptide Array |
Description
TRBP Monoclonal specifically detects TRBP in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.Specifications
| TRBP | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, Immunofluorescence, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Human | |
| LOQS, RISC-loading complex subunit TARBP2, TAR (HIV) RNA binding protein 2, TAR (HIV) RNA-binding protein 2, TAR (HIV) RNA-binding protein TRBP1, TAR (HIV-1) RNA binding protein 2, TAR RNA binding protein 2, TAR RNA-binding protein 2, trans-activation responsive RNA-binding protein, Trans-activation-responsive RNA-binding protein, TRBP, TRBP1, TRBP2 | |
| Recombinant full-length human TRBP protein | |
| Primary | |
| Store at 4°C. |
| PCRP-TARBP2-1E5 | |
| Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 6895 | |
| IgG2a | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title