missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRBP Mouse anti-Human, Clone: 1C9A10, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | TRBP |
|---|---|
| Clone | 1C9A10 |
| Concentration | 0.74 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16828995
|
Proteintech
66792-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16818995
|
Proteintech
66792-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRBP is an RNA associated nuclear protein. It contains 3 DRBM (double stranded RNA-binding) domains and plays an important role by binding to HIV-1 TAR RNA located in the long terminal repeat (LTR).TRBP acts synergistic with viral TAT protein in HIV-1 expression activation. It binds preferentially to TAR and other double-stranded RNAs with G+C-rich characteristics. It is important in regulation of transcription from RNA polymerase II promoter andis widely expressed in most of the tissues.Specifications
| TRBP | |
| 0.74 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| LOQS, TAR RNA binding protein 2, TARBP2, TRBP, TRBP1, TRBP2 | |
| TARBP2 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1C9A10 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q15633 | |
| 6895 | |
| TRBP Fusion Protein Ag8269 | |
| Primary | |
| -20°C | |
| TARBP2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title