missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ TRBC1 Antibody (5A8A10), Novus Biologicals™

Mouse Monoclonal Antibody

£226.00 - £389.00

Specifications

Antigen TRBC1
Clone 5A8A10
Concentration 1 mg/mL
Dilution Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000
Applications Flow Cytometry, ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30030855
View Documents
Novus Biologicals™
NBP3-27179-0.025ml
0.025 mL
£226.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30023560
View Documents
Novus Biologicals™
NBP3-27179-0.1ml
0.1 mL
£389.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TRBC1 Monoclonal antibody specifically detects TRBC1 in Human samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications

Specifications

TRBC1
1 mg/mL
Flow Cytometry, ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Adaptive Immunity, Cell Biology, Diabetes Research, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases
PBS
28639
IgG1
Protein G purified
5A8A10
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000
Monoclonal
Purified
RUO
Human
BV05S1J2.2, TCRBC1, TRBC1 T cell receptor beta constant 1
Purified recombinant fragment of human TRBC1 (AA: 1-149) expressed in E. Coli.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.