missing translation for 'onlineSavingsMsg'
Learn More

Trazodone Hydrochloride, MP Biomedicals™

Product Code. 11474772 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11474772 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11474772 Supplier MP Biomedicals Supplier No. 0219151801
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 25332-39-2
Molecular Formula C19H23Cl2N5O
Quantity 1 g
Synonym trazodone hydrochloride, trazodone hcl, desyrel, bimaran, molipaxin, devidon, pragmazone, thombran, tombran, tritico
InChI Key OHHDIOKRWWOXMT-UHFFFAOYSA-N
SMILES C1CN(CCN1CCCN2C(=O)N3C=CC=CC3=N2)C4=CC(=CC=C4)Cl.Cl
IUPAC Name 2-[3-[4-(3-chlorophenyl)piperazin-1-yl]propyl]-[1,2,4]triazolo[4,3-a]pyridin-3-one;hydrochloride
Molecular Weight (g/mol) 408.327
PubChem CID 62935
ChEBI CHEBI:9655
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.