missing translation for 'onlineSavingsMsg'
Learn More

Trazodone hydrochloride, MedChemExpress

Product Code. 16402156
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16402156

Brand: Medchem Express HYB0478

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Trazodone (hydrochloride) (AF-1161) is an antidepressant belonging to the class of serotonin receptor antagonists and reuptake inhibitors for treatment of anxiety disorders.

Trazodone (hydrochloride) (AF-1161) is an antidepressant belonging to the class of serotonin receptor antagonists and reuptake inhibitors for treatment of anxiety disorders.

Specifications

Chemical Name or Material Trazodone hydrochloride
Physical Form Solid
CAS 25332-39-2
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C19H23Cl2N5O
For Use With (Application) Neuroscience-Neuromodulation
Synonym AF-1161
Solubility Information DMSO : 16.67 mg/mL (40.83 mM; Need ultrasonic) ∣H2O : 16.67 mg/mL (40.83 mM; Need ultrasonic)
SMILES O=C1N(CCCN2CCN(C3=CC=CC(Cl)=C3)CC2)N=C4C=CC=CN41.Cl
Molecular Weight (g/mol) 408.32
Formula Weight 408.32
Percent Purity 99.87%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.