missing translation for 'onlineSavingsMsg'
Learn More

Trazodone hydrochloride, MedChemExpress

Product Code. 16418336
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16418336 100 mg 100mg
16402156 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16418336 Supplier Medchem Express Supplier No. HYB0478
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Trazodone (hydrochloride) (AF-1161) is an antidepressant belonging to the class of serotonin receptor antagonists and reuptake inhibitors for treatment of anxiety disorders.

Trazodone (hydrochloride) (AF-1161) is an antidepressant belonging to the class of serotonin receptor antagonists and reuptake inhibitors for treatment of anxiety disorders.

Specifications

Chemical Name or Material Trazodone hydrochloride
Physical Form Solid
CAS 25332-39-2
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C19H23Cl2N5O
For Use With (Application) Neuroscience-Neuromodulation
Synonym AF-1161
Solubility Information DMSO : 16.67 mg/mL (40.83 mM; Need ultrasonic) ∣H2O : 16.67 mg/mL (40.83 mM; Need ultrasonic)
SMILES O=C1N(CCCN2CCN(C3=CC=CC(Cl)=C3)CC2)N=C4C=CC=CN41.Cl
Molecular Weight (g/mol) 408.32
Formula Weight 408.32
Percent Purity 99.87%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.