missing translation for 'onlineSavingsMsg'
Learn More

Trazodone hydrochloride

Product Code. 18730949
Change view
Click to view available options
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
18730949 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18730949 Supplier Tocris Bioscience Supplier No. 6336/50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

5-HT2A and α1 adrenoceptor antagonist; also enhances neural differentiation; antidepressant and neuroprotectant

Trazodone hydrochloride is a 5-HT2A and α1 adrenoceptor antagonist. Enhances neuronal differentiation of mouse and human neural progenitor cells. Also inhibits PERK/eIF2α-P-mediated reduction in protein synthesis and restores memory deficits in a mouse dementia model. Antidepressant and neuroprotectant.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[3-[4-(3-Chlorophenyl)-1-piperazinyl]propyl]-1,2,4-triazolo[4,3-a]pyridin-3(2H)-one hydrochloride
CAS 25332-39-2
Assay Percent Range >99%
Quantity 50 mg
Molecular Formula C19H22ClN5O.HCl
Formula Weight Observed MW: 408.32
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.