missing translation for 'onlineSavingsMsg'
Learn More

Trazodone hydorchloride, 99.9%, MP Biomedicals™

Product Code. 11484772 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11484772 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11484772 Supplier MP Biomedicals Supplier No. 0219151805
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 25332-39-2
Color White
Assay Percent Range 0.999
Molecular Formula C19H23Cl2N5O
Quantity 5 g
Synonym trazodone hydrochloride, trazodone hcl, desyrel, bimaran, molipaxin, devidon, pragmazone, thombran, tombran, tritico
InChI Key OHHDIOKRWWOXMT-UHFFFAOYSA-N
SMILES C1CN(CCN1CCCN2C(=O)N3C=CC=CC3=N2)C4=CC(=CC=C4)Cl.Cl
IUPAC Name 2-[3-[4-(3-chlorophenyl)piperazin-1-yl]propyl]-[1,2,4]triazolo[4,3-a]pyridin-3-one;hydrochloride
Molecular Weight (g/mol) 408.327
PubChem CID 62935
ChEBI CHEBI:9655
Formula Weight 408.3
Physical Form Crystalline Powder
Chemical Name or Material Trazodone hydorchloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.