missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TRAX Monoclonal antibody specifically detects TRAX in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | TRAX |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | S02-3D3 |
| Conjugate | Unconjugated |
| Formulation | 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA |
| Gene Alias | DISC1, translin-associated factor Xdisrupted in schizophrenia 1, translin-like protein, TRAXtranslin-associated protein X |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide of human TRAX (Uniprot#: Q99598) |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?