missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Travoprost, MedChemExpress
Description
Travoprost (Fluprostenol isopropyl ester), an isopropyl ester prodrug, is a high affinity, selective FP prostaglandin full receptor agonist. Travoprost has the ocular hypotensive efficacy and has the potential for glaucoma and ocular hypertension.
Specifications
Specifications
| Chemical Name or Material | Travoprost |
| CAS | 157283-68-6 |
| Color | Colorless |
| Purity Grade Notes | Research |
| Molecular Formula | C26H35F3O6 |
| Quantity | 1 mg |
| Synonym | Fluprostenol isopropyl ester AL6221 Flu-Ipr |
| Solubility Information | Ethanol : 60 mg/mL (119.87 mM; Need ultrasonic) ∣DMSO : ≥ 41.67 mg/mL (83.25 mM) |
| SMILES | O=C(OC(C)C)CCC/C=C\C[C@@H]1[C@@H](/C=C/[C@@H](O)COC2=CC=CC(C(F)(F)F)=C2)[C@H](O)C[C@@H]1O |
| Molecular Weight (g/mol) | 500.55 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?