missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Trastuzumab

Product Code. 16463069
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16463069 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16463069 Supplier MedChemExpress Supplier No. HYP9907
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Trastuzumab is a humanized IgG1 monoclonal antibody for patients with invasive breast cancers that overexpress HER2. Trastuzumab has the potential for HER2 Positive Metastatic Breast Cancer and HER2 Positive Gastric Cancer research.

Specifications

Antigen HER2
Quantity 1 mg
Target Species Human
Content And Storage Please store the product under the recommended conditions in the Certificate of Analysis.
Product Type Inhibitory Antibodies
Shipping Condition Room temperature
CAS 180288-69-1
Purity or Quality Grade 99.2%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.