missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trappin-2/Elafin/Skalp Rabbit anti-Human, Clone: 101, Novus Biologicals™
Rabbit Monoclonal Antibody
£286.00 - £429.00
Specifications
| Antigen | Trappin-2/Elafin/Skalp |
|---|---|
| Clone | 101 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18253937
|
Novus Biologicals
NBP2-90071-50UL |
50 μg |
£286.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18286996
|
Novus Biologicals
NBP2-90071-100UL |
100 μg |
£429.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Trappin-2/Elafin/Skalp Monoclonal specifically detects Trappin-2/Elafin/Skalp in Human samples. It is validated for ELISA.Specifications
| Trappin-2/Elafin/Skalp | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 5266 | |
| IgG | |
| Protein A purified |
| 101 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| cementoin, Elastase-specific inhibitor, ESIWAP four-disulfide core domain protein 14, Peptidase inhibitor 3, peptidase inhibitor 3, skin-derived, PI-3, pre-elafin, protease inhibitor 3, skin-derived (SKALP), Protease inhibitor WAP3, SKALPELAFIN, Skin-derived antileukoproteinase, trappin-2, WAP four-disulfide core domain 14, WAP3MGC13613, WFDC14elafin | |
| Recombinant Human Trappin-2/Elafin/Skalp protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title