missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trappin-2/Elafin/Skalp Antibody (101), DyLight 550, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90071R
This item is not returnable.
View return policy
Description
Trappin-2/Elafin/Skalp Monoclonal specifically detects Trappin-2/Elafin/Skalp in Human samples. It is validated for ELISA.
Specifications
| Trappin-2/Elafin/Skalp | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| No cross-reactivity in ELISA with: Human SLPI Human TGM2 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 101 | |
| ELISA | |
| cementoin, Elastase-specific inhibitor, ESIWAP four-disulfide core domain protein 14, Peptidase inhibitor 3, peptidase inhibitor 3, skin-derived, PI-3, pre-elafin, protease inhibitor 3, skin-derived (SKALP), Protease inhibitor WAP3, SKALPELAFIN, Skin-derived antileukoproteinase, trappin-2, WAP four-disulfide core domain 14, WAP3MGC13613, WFDC14elafin | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Trappin-2/Elafin/Skalp (NP_002629.1; Met1-Gln117). | |
| 0.1 mL | |
| Cancer, Immunology | |
| 5266 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction