missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAPPC9 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93186-0.1ml
This item is not returnable.
View return policy
Description
TRAPPC9 Polyclonal antibody specifically detects TRAPPC9 in Mouse samples. It is validated for Western Blot
Specifications
| TRAPPC9 | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| IKBKBBP, IKK2 binding protein, KIAA1882IBP, MRT13MGC4769, NIBPMGC4737, NIK- and IKBKB-binding protein, NIK and IKK-beta binding protein, T1, trafficking protein particle complex 9, trafficking protein particle complex subunit 9, TRAPP 120 kDa subunit, TRS120, Tularik gene 1 protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 947-1246 of human TRAPPC9 (NP_113654.4). SHVKTLEAVLNFKYSGGPGHTEGYYRNLSLGLHVEVEPSVFFTRVSTLPATSTRQCHLLLDVFNSTEHELTVSTRSSEALILHAGECQRMAIQVDKFNFESFPESPGEKGQFANPKQLEEERREARGLEIHSKLGICWRIPSLKRSGEASVEGLLNQLVLEHLQLAPLQWDVLVDGQPCDREAVAACQVGDPVRLEVRLTNRSPRSVGPFALTVVPFQDHQNGVHNYDLHDTVSFVGSSTFYLDAVQPSGQSACLGALLFLYTGDFFLHIRFHEDSTSKELPPSWFCLPSVHVCALEAQA | |
| 0.1 mL | |
| Signal Transduction | |
| 83696 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction