missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRAPPC6B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561627
This item is not returnable.
View return policy
Description
Immunogen sequence: MADEALFLLL HNEMVSGVYK SAEQGEVENG RCITKLE Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 100%.
TRAPPC6B is a component of TRAPP complexes, which are tethering complexes involved in vesicle transport.
Specifications
| TRAPPC6B | |
| Polyclonal | |
| Unconjugated | |
| TRAPPC6B | |
| 5830498C14Rik; C79212; RGD1309325; TPC6; trafficking protein particle complex 6b; trafficking protein particle complex subunit 6b; Trafficking protein particle complex subunit 6B-like protein; TRAPP complex subunit 6B; Trappc6b; Unknown (protein for MGC:133693); zgc:103464 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 122553 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86SZ2 | |
| TRAPPC6B | |
| Recombinant protein corresponding to Human TRAPPC6B. Recombinant protein control fragment (Product #RP-100490). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction