missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAPPC5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA568346
This item is not returnable.
View return policy
Description
Predicted to react with Mouse and Rat samples.
May play a role in vesicular transport from endoplasmic reticulum to Golgi.
Specifications
| TRAPPC5 | |
| Polyclonal | |
| Unconjugated | |
| TRAPPC5 | |
| TRAPPC5 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 126003, 363858, 66682 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9CQA1, Q8IUR0 | |
| Trappc5 | |
| Synthesized peptide derived from human TRAPPC5. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction