missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRAPPC5 Monoclonal Antibody (3D9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524512
This item is not returnable.
View return policy
Description
Peptide Sequence: MEARFTRGKS ALLERALARP RTEVSLSAFA LLFSELVQHC QSRVFSVAEL QSRLAALGRQ VGARVLDALV AREKGARRET KVLGALLFVK GAVWKALFGK EADKLEQAND DARTFYIIER EPLINTYISV PKENSTLNCA SFTAGIVEAV LTHSGFPAKV TAHWHKGTTL MIKFEEAVIA RDRALEGR.
TRAPPC5 (trafficking protein particle complex 5), also known as TRS31, is a 181 amino acid protein belonging to the TRAPP (transport protein particle) small subunits family and the BET3 subfamily. Encoded by a gene that maps to human chromosome 19p13.2, TRAPPC5 is part of the multisubunit TRAPP tethering complex, which acts as a GTP exchange factor. Evolutionarily conserved, TRAPPC5 plays a role in protein binding,vesicle-mediated transport and nucleotide exchange stimulation. TRAPPC5 also performs guanine nucleotide exchanger factor (GEF) functions both in vitro and in vivo. Localizing to the Golgi apparatus, TRAPPC5 is essential for endoplasmic reticulum (ER)-to-Golgi and intra-Golgi vesicle trafficking in yeast, as well as additional transport events in mammals, such as post-Golgi trafficking.
Specifications
| TRAPPC5 | |
| Monoclonal | |
| Unconjugated | |
| TRAPPC5 | |
| 4021401A16Rik; RGD1563971; trafficking protein particle complex 5; trafficking protein particle complex subunit 5; Trappc5; TRS31 | |
| Mouse | |
| Protein A | |
| RUO | |
| 126003 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA | |
| 3D9 | |
| PBS with no preservative; pH 7.4 | |
| Q8IUR0 | |
| TRAPPC5 | |
| TRAPPC5 (AAH42161, 1 a.a. ~ 188 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction