missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAPPC5 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18481IR
This item is not returnable.
View return policy
Description
TRAPPC5 Polyclonal antibody specifically detects TRAPPC5 in Human, Mouse samples. It is validated for Western Blot
Specifications
| TRAPPC5 | |
| Polyclonal | |
| Western Blot | |
| MGC52424, trafficking protein particle complex 5, trafficking protein particle complex subunit 5, TRS31 | |
| Recognizes region between residues Between between 138 and 188.. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 126003 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction