missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAPPC4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12074-1-AP-150UL
This item is not returnable.
View return policy
Description
TRAPPC4 is part of the multisubunit TRAPP (transport protein particle) complex and interacts with SDC2. It may play a role in vesicular transport from endoplasmic reticulum to Golgi. TRAPP proteins are involved in tethering during vesicle transport.Specifications
| TRAPPC4 | |
| Polyclonal | |
| Unconjugated | |
| TRAPPC4 | |
| CGI 104, HSPC172, PTD009, SBDN, SYNBINDIN, TRAPPC4, TRS23, TRS23 homolog | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 367073, 51399, 60409 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q69BT7, Q9ES56, Q9Y296 | |
| TRAPPC4 | |
| TRAPPC4 Fusion Protein Ag2737 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction