missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRAPPC4 Monoclonal Antibody (OTI2E1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526162
This item is not returnable.
View return policy
Description
TRAPPC4 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
TRAPPC4 is part of the multisubunit TRAPP (transport protein particle) complex and interacts with SDC2. It may play a role in vesicular transport from endoplasmic reticulum to Golgi. TRAPP proteins are involved in tethering during vesicle transport.
Specifications
| TRAPPC4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9Y296 | |
| Trappc4 | |
| Full length human recombinant protein of TRAPPC4 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI2E1 | |
| Unconjugated | |
| Trappc4 | |
| 1500017G03Rik; AI303617; CGI-104; Hematopoietic stem/progenitor cell protein 172; HSPC172; hypothetical protein LOC504391; PTD009; Sbd; Sbdn; Sdcbp2; synbindin; syndecan binding protein 2; trafficking protein particle complex 4; trafficking protein particle complex subunit 4; Trappc4; TRS23; TRS23 homolog; zgc:73260 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 51399 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction