missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRAPPC2 Recombinant Rabbit Monoclonal Antibody (K01_2P83)

Product Code. 30829629
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30829629 50 μL
30825563 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30829629 Supplier Invitrogen™ Supplier No. MA565684
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

TRAPPC2 Recombinant Monoclonal Antibody for Western Blot

The protein encoded by this gene is thought to be part of a large multi-subunit complex involved in the targeting and fusion of endoplasmic reticulum-to-Golgi transport vesicles with their acceptor compartment. In addition, the encoded protein can bind c-myc promoter-binding protein 1 and block its transcriptional repression capability. Mutations in this gene are a cause of spondyloepiphyseal dysplasia tarda (SEDT). A processed pseudogene of this gene is located on chromosome 19, and other pseudogenes are found on chromosomes 8 and Y. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAPPC2
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_2P83
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene TRAPPC2
Gene Accession No. P0DI81
Gene Alias 1110066L09Rik; 1810064C02Rik; AW496358; hYP38334; hypothetical protein LOC616748; MBP-1-interacting protein 2A; MIP2A; MIP-2A; RGD1306925; SEDL; Sedlin; SEDLP1; SEDT; spondyloepiphyseal dysplasia tarda protein; spondyloepiphyseal dysplasia, late; trafficking protein particle complex 2; trafficking protein particle complex protein 2; Trafficking protein particle complex subunit 2; Trafficking protein particle complex subunit 2B; Trappc2; TRAPPC2.19; TRAPPC2B; TRAPPC2P1; TRS20; zgc:152903; ZNF547L
Gene Symbols TRAPPC2
Host Species Rabbit
Immunogen A synthetic peptide of human TRAPPC2.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6399
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.