missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRAPPC13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558098
This item is not returnable.
View return policy
Description
Immunogen sequence: LGETFSSYIS VHNDSNQVVK DILVKADLQT SSQRLNLSAS NAAVAELKPD CCIDDVIHHE VKEIGTHILV CAVSY Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
The function of C5orf44 has not been widely studied, and is yet to be fully elucidated.
Specifications
| TRAPPC13 | |
| Polyclonal | |
| Unconjugated | |
| Trappc13 | |
| 2410002O22Rik; 2610524F24Rik; AI451892; AU017229; C5orf44; RGD1306583; trafficking protein particle complex 13; trafficking protein particle complex subunit 13; TRAPPC13; Trs65-related; UPF0533 protein C5orf44; UPF0533 protein C5orf44 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 80006 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A5PLN9 | |
| Trappc13 | |
| Recombinant protein corresponding to Human TRAPPC13. Recombinant protein control fragment (Product #RP-96832). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction