Learn More
TRAPPC10 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27549-1-AP-150UL
Description
The TRAPPC10 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TRAPPC10. This antibody recognizes human, mouse, rat antigen. The TRAPPC10 antibody has been validated for the following applications: IHC, IP, ELISA analysis.
Specifications
| TRAPPC10 | |
| 130 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA | |
| EHOC 1, EHOC1, GT334, Protein GT334, TMEM1, TRAPP subunit TMEM1, TRAPPC10, TRS130, TRS30 | |
| 150 μL | |
| 7109 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |