missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRAPPC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568345
This item is not returnable.
View return policy
Description
Predicted to react with Human, Mouse, and Rat samples.
TRAPPC1 plays a role in vesicular transport of proteins to the Golgi apparatus from the endoplasmic reticulum. The encoded protein is a component of the multisubunit transport protein particle (TRAPP) complex. Alternative splicing results in multiple transcript variants.
Specifications
| TRAPPC1 | |
| Polyclonal | |
| Unconjugated | |
| TRAPPC1 | |
| BET5; BET5 homolog; BET5-like protein; D11Ertd172e; EGK_08154; melanoma ubiquitous mutated 2; multiple myeloma protein 2; MUM2; MUM-2; novel Sybindin-like family domain-containing protein; trafficking protein particle complex 1; trafficking protein particle complex subunit 1; TRAPPC1; zgc:100969 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 245828, 287427, 58485 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q2KMM2, Q5NCF2, Q9Y5R8 | |
| TRAPPC1 | |
| Synthesized peptide derived from human TRAPPC1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction