missing translation for 'onlineSavingsMsg'
Learn More

Trapidil EP Impurity A, Mikromol™

Product Code. 30050799
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050799 25 mg 25mg
30049705 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30050799 Supplier Mikromol Supplier No. MM3502.010025
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Trapidil EP Impurity A
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-methyl-[1,2,4]triazolo[1,5-a]pyrimidin-7-ol
CAS 2503-56-2
Analyte or Component Names 5-methyl-[1,2,4]triazolo[1,5-a]pyrimidin-7-ol
Shipping Condition Room Temperature
Molecular Formula C6 H6 N4 O
Certifications/Compliance ISO 17025
Synonym 5-Methyl-[1,2,4]triazolo[1,5-a]pyrimidin-7-ol, Trapidil Imp. A (EP), 5-Methyl-[1,2,4]triazolo[1,5-a]pyrimidin-7-ol, 5-Methyl-s-triazolo[1,5-a]pyrimidin-7-ol, 1,3,7-Triaza-4-hydroxy-6-methylindolizine, 4-Hydroxy-6-methyl-1,2,4-triazolo[2,3-a]pyrimidine, 4-Hydroxy-6-methyl-1,3,3a,7-tetraazaindene, 4-Hydroxy-6-methyl-1,3,3a,7-tetrazaindene, 5-Methyl-7-hydroxy-1,2,4-triazolo[1,5-a]pyrimidine, 5-Methyl-7-hydroxy-1,3,4-triazaindolizine, 5-Methyl-7-hydroxy-s-triazolo[1,5-a]pyrimidine, 6-Methyl-4-hydroxy-1,3,3a,7-tetraazaindene, 7-Hydroxy-5-methyl-1,3,4-triazaindolizine, 7-Hydroxy-5-methyl-s-triazolo[1,5-a]pyrimidine, NSC 32071, NSC 511493, Stabilizer 583, TAI, TAI (sensitizer), Trapidil Impurity A
InChI Formula InChI=1S/C6H6N4O/c1-4-2-5(11)10-6(9-4)7-3-8-10/h2-3,11H,1H3
SMILES Cc1cc(O)n2ncnc2n1
IUPAC Name 5-methyl-[1,2,4]triazolo[1,5-a]pyrimidin-7-ol
Molecular Weight (g/mol) 150.14
Quantity 25 mg
Formula Weight 150.0542 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.