Learn More
TRAPA/SSR1 Mouse anti-Human, Unconjugated, Clone: 1F2D7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67925-1-IG-150UL
Description
The TRAPA/SSR1 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human TRAPA/SSR1. This antibody recognizes human, mouse, rat antigen. The TRAPA/SSR1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| TRAPA/SSR1 | |
| Monoclonal | |
| Unconjugated | |
| 150 μL | |
| 6745 | |
| IgG1 |
| Western Blot, Immunohistochemistry, ELISA | |
| 1F2D7 | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat |