missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRAP1 Recombinant Rabbit Monoclonal Antibody (24GB2560)

Rabbit Recombinant Monoclonal Antibody
£172.00 - £488.00
Specifications
| Antigen | TRAP1 |
|---|---|
| Clone | 24GB2560 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30275762
![]() |
Invitrogen™
MA557786 |
100 μL |
£488.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275971
|
Invitrogen™
MA557787 |
20 μL |
£172.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRAP1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The 90 kDa heat shock protein (hsp90) family of molecular chaperones is a highly conserved family of proteins that play an important physiological role. Hsp90 is involved in numerous cellular processes but is best known for its association with signal transduction machinery. A recently cloned homolog of hsp90 is the tumor necrosis factor receptor-associated protein (TRAP1). Like hsp90, TRAP1 is found to be associated with numerous proteins involved in diverse actions. Immunofluorescence data has shown TRAP1 to be localized in the mitochondria of mammalian cells. This observation and the fact that TRAP1 is shown to have a mitochondrial targeting presequence strongly implicates TRAP1 as a mitochondrial matrix protein.Specifications
| TRAP1 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| Trap1 | |
| 2410002K23Rik; heat shock protein 75 kDa, mitochondrial; HSP 75; HSP75; HSP90L; testicular tissue protein Li 209; TNF receptor associated protein 1; TNF receptor-associated protein 1; TNFR-associated protein 1; TRAP1; TRAP-1; tumor necrosis factor type 1 receptor associated protein; tumor necrosis factor type 1 receptor-associated protein | |
| Trap1 | |
| Primary | |
| Affinity chromatography |
| 24GB2560 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q12931 | |
| 10131 | |
| A synthesized peptide derived from human TRAP1 (aa 50-100). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title