missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TRAP1 Monoclonal antibody specifically detects TRAP1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation
Specifications
Specifications
| Antigen | TRAP1 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | 1L8C1 |
| Conjugate | Unconjugated |
| Formulation | PBS, 0.05% BSA, 50% glycerol, pH7.3 |
| Gene Alias | HSP 75, HSP75TNFR-associated protein 1, HSP90Lheat shock protein 75 kDa, mitochondrial, TNF receptor-associated protein 1, TRAP-1, tumor necrosis factor type 1 receptor associated protein, Tumor necrosis factor type 1 receptor-associated protein |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human TRAP1 (Q12931). |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?