missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAP alpha Antibody (OTI4C7), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-74591CL1
This item is not returnable.
View return policy
Description
TRAP alpha Monoclonal antibody specifically detects TRAP alpha in Canine,Human,Monkey,Mouse,Primate,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF,Immunocytochemistry/Immunofluorescence
Specifications
| TRAP alpha | |
| Monoclonal | |
| CoraFluor 1 | |
| DKFZp781N23103, FLJ14232, FLJ22100, FLJ23034, FLJ78242, FLJ93042, signal sequence receptor, alpha, SSR alpha subunit, SSR-alpha, translocon-associated protein alpha subunit, translocon-associated protein subunit alpha, TRAP alpha, TRAP-alpha, TRAPASignal sequence receptor subunit alpha | |
| Full length human recombinant protein of human SSR1(NP_003135) produced in HEK293 cell. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 6745 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF, Immunocytochemistry/Immunofluorescence | |
| OTI4C7 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Canine, Human, Monkey, Mouse, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction