missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TRAP alpha Polyclonal antibody specifically detects TRAP alpha in Human,Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | TRAP alpha |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Tris-citrate/phosphate pH 7 -8. |
| Gene Alias | DKFZp781N23103, FLJ14232, FLJ22100, FLJ23034, FLJ78242, FLJ93042, signal sequence receptor, alpha, SSR alpha subunit, SSR-alpha, translocon-associated protein alpha subunit, translocon-associated protein subunit alpha, TRAP alpha, TRAP-alpha, TRAPASignal sequence receptor subunit alpha |
| Host Species | Rabbit |
| Immunogen | Between 236 and 286 |
| Purification Method | Affinity purified |
| Quantity | 100 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?