missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transthyretin/Prealbumin Mouse anti-Human, Clone: TTR/4292, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07742-100ug
This item is not returnable.
View return policy
Description
Transthyretin/Prealbumin Monoclonal specifically detects Transthyretin/Prealbumin in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| Transthyretin/Prealbumin | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 2-4 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array | |
| ATTR, HsT2651, PALBTBPA, Prealbumin, prealbumin, amyloidosis type I, thyroxine-binding prealbumin, transthyretin | |
| Recombinant human full-length protein | |
| 100 μg | |
| Cancer | |
| 7276 | |
| Store at 4°C. | |
| IgG2c κ |
| Western Blot, Immunohistochemistry (Paraffin), Peptide Array | |
| TTR/4292 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction