missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transthyretin/Prealbumin Antibody (6F11B2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£438.00
Specifications
| Antigen | Transthyretin/Prealbumin |
|---|---|
| Clone | 6F11B2 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry |
Description
Transthyretin/Prealbumin Monoclonal specifically detects Transthyretin/Prealbumin in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
| Transthyretin/Prealbumin | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| PBS with 0.05% Sodium Azide | |
| 7276 | |
| Purified recombinant fragment of human TTR (AA: 1-147) expressed in E. Coli. | |
| Primary | |
| 16 kDa |
| 6F11B2 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ATTR, HsT2651, PALBTBPA, Prealbumin, prealbumin, amyloidosis type I, thyroxine-binding prealbumin, transthyretin | |
| TTR | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title