missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transthyretin/Prealbumin Antibody (003), DyLight 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90050DL594
This item is not returnable.
View return policy
Description
Transthyretin/Prealbumin Monoclonal specifically detects Transthyretin/Prealbumin in Human samples. It is validated for ELISA.
Specifications
| Transthyretin/Prealbumin | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 3 | |
| ELISA | |
| ATTR, HsT2651, PALBTBPA, Prealbumin, prealbumin, amyloidosis type I, thyroxine-binding prealbumin, transthyretin | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Transthyretin/Prealbumin (NP_000362.1; Met 1-Glu 147). | |
| 0.1 mL | |
| Cancer | |
| 7276 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction