missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transportin-1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20679-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody is specific to TNPO1.
TNPO1 comprises the beta subunit of the karyopherin receptor complex which interacts with nuclear localization signals to target nuclear proteins to the nucleus. The karyopherin receptor complex is a heterodimer of an alpha subunit which recognizes the nuclear localization signal and a beta subunit which docks the complex at nucleoporins.Specifications
| Transportin-1 | |
| Polyclonal | |
| Unconjugated | |
| TNPO1 | |
| TNPO1, transportin 1, Transportin1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 238799, 3842 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8BFY9, Q92973 | |
| TNPO1 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction