missing translation for 'onlineSavingsMsg'
Learn More

Transplatin, TRC

Product Code. 30456418
Change view
Click to view available options
Quantity:
10 mg
100 mg
250 mg
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30456418 100 mg -
30452605 10 mg -
30449367 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456418 Supplier Toronto Research Chemicals Supplier No. TRCT714800100MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14913-33-8
Molecular Formula Cl2 H6 N2 Pt
Molecular Weight (g/mol) 300.04
Synonym Transplatin, (SP-4-1)-Diamminedichloro-platinum, trans-Diamminedichloro-platinum, NSC 131558, trans-DDP, trans-Diaminedichloroplatinum, trans-Diamminedichloroplatinum, trans-Diamminedichloroplatinum(II), trans-Dichlorodiamineplatinum(II), trans-Dichlorodiammine platinum, trans-Dichlorodiammineplatinum(II), trans-Platinum(II) ammonium chloride, trans-Platinumdiammine dichloride
SMILES [Cl-][Pt+2]([Cl-])([NH3])[NH3]

Specifications

Quantity 100 mg
InChI Formula InChI=1S/2ClH.2H3N.Pt/h2*1H;2*1H3;/q;;;;+2/p-2
Formula Weight 298.9556
Chemical Name or Material Transplatin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.