missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Deutsch Neumann™ Silicone Rubber Stoppers
For safe and secure closure of laboratory apparatus. Deutsch Neumann™ Silicone Rubber Stoppers are transparent in color.
Brand: Deutsch Neumann™ 1010508
Additional Details : Weight : 0.00090kg
Description
- Acc. to DIN 12 871
- Heat resistant up to 200°C
- Durometer 50 Shore A
- Available in a wide range of sizes
Specifications
Transparent | |
20mm | |
12 mm | |
Silicone |
Stopper | |
8 mm | |
1/Ea. |