missing translation for 'onlineSavingsMsg'
Learn More

BrandTech™ VITALAB™ Polypropylene Sample Vials with Snap-On Caps

These Polypropylene Sample Vials with Snap-On Caps are perfectly suited for the collection, storage and transport of samples in the laboratory.

Brand:  BrandTech™ 130494

66.68 GBP valid until 2024-12-31
Use promo code "23277" to get your promotional price.



Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10366722

  • £82.25 / Pack of 10
Estimated Shipment: 05-06-2024
to see stock.
Add to basket

Description

Description

  • Transparent in color
  • Snap caps and vial body from PP
  • Excellent resistance to alcohols, aldehydes, alkalis and acids
Specifications

Specifications

Transparent
Polypropylene
10
Polypropylene
62 mm
GL 56
54 mm
90 mL
125°C Max.
Screw Cap
Sample Container
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers